Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244208.1 | NP_001231137.1 | 1 - 2 | 290 | 88 | |
GenBank mRNA | AK238807.1 | - | 1 - 2 | 288 | 88 | |
AK351591.1 | - | 1 - 2 | 255 | 88 | ||
AK351936.1 | - | 1 - 2 | 248 | 88 | ||
AK392733.1 | - | 1 - 2 | 263 | 88 | ||
AK396232.1 | - | 1 - 2 | 248 | 88 | ||
AK397156.1 | - | 1 - 2 | 253 | 88 | ||
AK398185.1 | - | 1 - 2 | 248 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |