Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244208.1 | NP_001231137.1 | 4 - 5 | 718 | 92 | |
GenBank mRNA | AK238807.1 | - | 4 - 5 | 716 | 92 | |
AK351591.1 | - | 4 - 5 | 683 | 92 | ||
AK351936.1 | - | 4 - 5 | 676 | 92 | ||
AK392733.1 | - | 4 - 5 | 691 | 92 | ||
AK396232.1 | - | 4 - 5 | 676 | 92 | ||
AK397156.1 | - | 4 - 5 | 681 | 92 | ||
AK397840.1 | - | 4 - 5 | 718 | 92 | ||
AK398185.1 | - | 4 - 5 | 676 | 92 | ||
CJ022605.1 | - | 4 - 5 | 718 | 92 | ||
DB817925.1 | - | 1 - 2 | 553 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |