Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348754.1 | NP_001335683.1 | 1 - 2 | 523 | 143 |
Gene Symbol: LOC100622780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK350081.1 | - | 1 - 2 | 524 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |