Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244241.1 | NP_001231170.1 | 4 - 5 | 340 | 97 | |
NM_001244242.1 | NP_001231171.1 | 4 - 5 | 324 | 97 | ||
XM_005659114.3 | XP_005659171.1 | 359 | 97 | |||
XM_021101793.1 | XP_020957452.1 | 359 | 97 | |||
XM_021101851.1 | XP_020957510.1 | 481 | 97 | |||
GenBank mRNA | AK231140.1 | - | 5 - 6 | 224 | 97 | |
AK233198.1 | - | 4 - 5 | 314 | 97 | ||
AK236344.1 | - | 4 - 5 | 324 | 97 | ||
AK237427.1 | - | 5 - 6 | 221 | 97 | ||
AK351349.1 | - | 4 - 5 | 340 | 97 | ||
AK391167.1 | - | 4 - 5 | 324 | 97 | ||
AK394413.1 | - | 4 - 5 | 311 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |