Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244261.1 | NP_001231190.1 | 6 - 7 | 899 | 82 | |
GenBank mRNA | HQ630067.1 | - | 6 - 7 | 919 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |