Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244280.2 | NP_001231209.1 | 10 - 11 | 1343 | 73 | |
XM_021093374.1 | XP_020949033.1 | 1827 | 73 | |||
GenBank mRNA | HQ654522.1 | - | 10 - 11 | 1344 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |