Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244280.2 | NP_001231209.1 | 6 - 7 | 655 | 66 | |
XM_021093374.1 | XP_020949033.1 | 812 | 66 | |||
GenBank mRNA | HQ654522.1 | - | 6 - 7 | 656 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |