Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244284.1 | NP_001231213.1 | 7 - 8 | 972 | 56 |
Gene Symbol: PGLYRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB292178.1 | - | 7 - 8 | 972 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |