Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244288.1 | NP_001231217.1 | 1 - 2 | 326 | 89 | |
NM_001244289.1 | NP_001231218.1 | 2 - 3 | 374 | 89 | ||
GenBank mRNA | AK233501.1 | - | 2 - 3 | 374 | 89 | |
AK234190.1 | - | 1 - 2 | 326 | 89 | ||
AK238776.1 | - | 2 - 3 | 233 | 89 | ||
AK395093.1 | - | 1 - 2 | 326 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |