Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244291.1 | NP_001231220.1 | 1 - 2 | 840 | 86 | |
XM_005662003.3 | XP_005662060.1 | 1189 | 86 | |||
XM_013995569.2 | XP_013851023.1 | 1163 | 86 | |||
XM_021085683.1 | XP_020941342.1 | 1014 | 86 | |||
XM_021085684.1 | XP_020941343.1 | 1037 | 86 | |||
GenBank mRNA | HQ439987.1 | - | 1 - 2 | 840 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |