Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244329.1 | NP_001231258.1 | 3 - 4 | 498 | 108 | |
XM_005669062.3 | XP_005669119.2 | 523 | 108 | |||
XM_021066135.1 | XP_020921794.1 | 526 | 108 | |||
GenBank mRNA | AK343440.1 | - | 3 - 4 | 498 | 108 | |
AK351417.1 | - | 3 - 4 | 429 | 108 | ||
DB787462.1 | - | 3 - 4 | 498 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |