Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244329.1 | NP_001231258.1 | 7 - 8 | 1139 | 74 | |
XM_021066135.1 | XP_020921794.1 | 1167 | 74 | |||
GenBank mRNA | AK343440.1 | - | 7 - 8 | 1139 | 74 | |
CJ019492.1 | - | 4 - 5 | 587 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |