Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIBAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244334.1 | NP_001231263.1 | 1 - 2 | 119 | 65 | |
XM_021093319.1 | XP_020948978.1 | 903 | 65 | |||
GenBank mRNA | AJ684804.1 | - | 1 - 2 | 86 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |