Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIBAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244334.1 | NP_001231263.1 | 6 - 7 | 634 | 81 | |
XM_021093319.1 | XP_020948978.1 | 1418 | 81 | |||
XM_021093320.1 | XP_020948979.1 | 1542 | 81 | |||
GenBank mRNA | AK349407.1 | - | 6 - 7 | 634 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |