Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244361.1 | NP_001231290.1 | 2 - 3 | 430 | 108 | |
GenBank mRNA | AB295597.2 | - | 1 - 2 | 85 | 108 | |
AK397291.1 | - | 2 - 3 | 443 | 108 | ||
DB802979.1 | - | 2 - 3 | 430 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |