Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244377.1 | NP_001231306.1 | 1 - 2 | 377 | 96 | |
GenBank mRNA | EW394041.2 | - | 1 - 2 | 179 | 96 | |
EW493122.2 | - | 1 - 2 | 282 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |