Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244386.1 | NP_001231315.1 | 9 - 10 | 1108 | 89 | |
XM_021100359.1 | XP_020956018.1 | 1707 | 89 | |||
GenBank mRNA | AY609657.1 | - | 9 - 10 | 1108 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |