Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244387.1 | NP_001231316.1 | 3 - 4 | 347 | 74 | |
XM_021088495.1 | XP_020944154.1 | 792 | 74 | |||
XM_021088496.1 | XP_020944155.1 | 779 | 74 | |||
XM_021088497.1 | XP_020944156.1 | 733 | 74 | |||
XM_021088499.1 | XP_020944158.1 | 313 | 74 | |||
XM_021088500.1 | XP_020944159.1 | 468 | 74 | |||
XM_021088501.1 | XP_020944160.1 | 467 | 74 | |||
XM_021088502.1 | XP_020944161.1 | 729 | 74 | |||
GenBank mRNA | AK237145.1 | - | 3 - 4 | 338 | 74 | |
AK350371.1 | - | 3 - 4 | 324 | 74 | ||
AK396598.1 | - | 3 - 4 | 370 | 74 | ||
AY610035.1 | - | 3 - 4 | 347 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |