Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244400.1 | NP_001231329.1 | 7 - 8 | 900 | 101 | |
XM_005654539.3 | XP_005654596.1 | 1247 | 101 | |||
XM_005654541.3 | XP_005654598.1 | 963 | 101 | |||
XM_005654543.3 | XP_005654600.1 | 778 | 101 | |||
XM_013986596.2 | XP_013842050.1 | 882 | 101 | |||
GenBank mRNA | AK240368.1 | - | 7 - 8 | 900 | 101 | |
AK389388.1 | - | 7 - 8 | 900 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |