Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244414.2 | NP_001231343.2 | 5 - 6 | 473 | 70 | |
XM_021088484.1 | XP_020944143.1 | 438 | 70 | |||
XM_021088485.1 | XP_020944144.1 | 1226 | 70 | |||
XM_021088486.1 | XP_020944145.1 | 587 | 70 | |||
XM_021088487.1 | XP_020944146.1 | 549 | 70 | |||
XM_021088488.1 | XP_020944147.1 | 558 | 70 | |||
XM_021088490.1 | XP_020944149.1 | 555 | 70 | |||
GenBank mRNA | AK230648.1 | - | 5 - 6 | 441 | 70 | |
AK230826.1 | - | 5 - 6 | 458 | 70 | ||
AK230913.1 | - | 5 - 6 | 458 | 70 | ||
AK230930.1 | - | 5 - 6 | 456 | 70 | ||
AK230937.1 | - | 5 - 6 | 460 | 70 | ||
AK343772.1 | - | 5 - 6 | 473 | 70 | ||
AK347640.1 | - | 11 - 12 | 1226 | 70 | ||
AK389623.1 | - | 5 - 6 | 458 | 70 | ||
AK389775.1 | - | 5 - 6 | 441 | 70 | ||
AK389788.1 | - | 5 - 6 | 472 | 70 | ||
AK389799.1 | - | 5 - 6 | 473 | 70 | ||
AK389825.1 | - | 5 - 6 | 592 | 70 | ||
AK391408.1 | - | 5 - 6 | 476 | 70 | ||
AK399467.1 | - | 5 - 6 | 441 | 70 | ||
AK399483.1 | - | 5 - 6 | 461 | 70 | ||
AY609570.1 | - | 5 - 6 | 509 | 70 |
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK389643.1 | - | 445 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |