Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244416.1 | NP_001231345.1 | 1 - 2 | 327 | 57 | |
GenBank mRNA | AK347068.1 | - | 1 - 2 | 298 | 57 | |
AK398591.1 | - | 1 - 2 | 286 | 57 | ||
AY610329.1 | - | 1 - 2 | 274 | 57 | ||
FD598500.1 | - | 1 - 2 | 327 | 57 | ||
FS699491.1 | - | 1 - 2 | 256 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |