Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244472.1 | NP_001231401.1 | 1 - 2 | 125 | 67 | |
GenBank mRNA | AK234065.1 | - | 1 - 2 | 125 | 67 | |
AK350385.1 | - | 1 - 2 | 52 | 67 | ||
AK389550.1 | - | 1 - 2 | 78 | 67 | ||
AK395923.1 | - | 1 - 2 | 136 | 67 | ||
AK398152.1 | - | 1 - 2 | 66 | 67 | ||
AK398286.1 | - | 1 - 2 | 62 | 67 | ||
AK399220.1 | - | 1 - 2 | 64 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |