Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244556.1 | NP_001231485.1 | 1 - 2 | 274 | 72 | |
GenBank mRNA | AJ667530.1 | - | 1 - 2 | 245 | 72 | |
CJ008617.1 | - | 1 - 2 | 261 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |