Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244622.1 | NP_001231551.1 | 2 - 3 | 588 | 89 | |
GenBank mRNA | AK349825.1 | - | 2 - 3 | 588 | 89 | |
AK397166.1 | - | 2 - 3 | 591 | 89 | ||
AK397400.1 | - | 2 - 3 | 585 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |