Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244624.1 | NP_001231553.1 | 10 - 11 | 951 | 66 | |
GenBank mRNA | AK232367.1 | - | 10 - 11 | 951 | 66 | |
AK394444.1 | - | 10 - 11 | 954 | 66 | ||
AK397989.1 | - | 10 - 11 | 941 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |