Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244626.1 | NP_001231555.1 | 5 - 6 | 778 | 107 | |
GenBank mRNA | AK345033.1 | - | 5 - 6 | 790 | 107 | |
AK391910.1 | - | 5 - 6 | 788 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |