Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244785.1 | NP_001231714.1 | 2 - 3 | 509 | 85 | |
XM_021078481.1 | XP_020934140.1 | 519 | 85 | |||
GenBank mRNA | AK347659.1 | - | 2 - 3 | 517 | 85 | |
AY610453.1 | - | 2 - 3 | 355 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |