Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244786.1 | NP_001231715.1 | 5 - 6 | 796 | 125 | |
GenBank mRNA | AK231845.1 | - | 5 - 6 | 798 | 125 | |
AY610052.1 | - | 5 - 6 | 785 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |