Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244801.1 | NP_001231730.1 | 3 - 4 | 373 | 91 | |
XM_013987749.2 | XP_013843203.1 | 91 | ||||
GenBank mRNA | AK239101.1 | - | 3 - 4 | 373 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |