Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244801.1 | NP_001231730.1 | 6 - 7 | 705 | 99 | |
XM_005655129.3 | XP_005655186.1 | 817 | 99 | |||
XM_013987749.2 | XP_013843203.1 | 847 | 99 | |||
GenBank mRNA | AK239101.1 | - | 6 - 7 | 705 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |