Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244124.1 | NP_001231053.1 | 1720 | 108 | ||
GenBank mRNA | AK232301.1 | - | 6 | 1715 | 108 | |
AK393957.1 | - | 6 | 1719 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |