Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCoAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB158406.1 | - | 44 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Triticum aestivum |