Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X89827.2 | - | 100 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Triticum aestivum |