Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF274001.1 | - | 450 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Triticum aestivum |