Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ878510.1 | - | 747 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Triticum aestivum |