Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY781360.1 | - | 646 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Triticum aestivum |