Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVVSPMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB240539.1 | - | 1 - 2 | 65 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |