Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VVSUC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF021808.1 | - | 3 - 4 | 1327 | 79 | |
AF182445.1 | - | 3 - 4 | 1297 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |