Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VVSUC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF021809.1 | - | 13 - 14 | 1683 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Vitis vinifera Probe spans exons |