Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VVSUC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF021809.1 | - | 3 - 4 | 470 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |