Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF373603.1 | - | 3 - 4 | 428 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Vitis vinifera Probe spans exons |