Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF373603.1 | - | 7 - 8 | 754 | 59 | |
AY256860.1 | - | 6 - 7 | 412 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |