Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ865334.1 | - | 2 - 3 | 446 | 65 | |
DQ129686.1 | - | 2 - 3 | 370 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |