Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY222741.1 | - | 1 - 2 | 40 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |