Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY233206.1 | - | 3 - 4 | 300 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |