Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VFUL-L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY275713.1 | - | 2 - 3 | 242 | 100 | |
AY538747.1 | - | 2 - 3 | 271 | 100 | ||
GU133635.1 | - | 1 - 2 | 188 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Vitis vinifera Probe spans exons |