Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY461710.1 | - | 8 - 9 | 842 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons |