Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY538259.1 | - | 3 - 4 | 1173 | 99 | |
AY854146.1 | - | 3 - 4 | 1247 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |