Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ504309.1 | - | 5 - 6 | 791 | 76 | |
GU133633.1 | - | 5 - 6 | 474 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |