Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF636725.2 | - | 1 | 1654 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Both primers and probe map within a single exon |